ENA638587232 Enamine

ethyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxy-3-(pyridin-4-yl)propanoate

  • MDL number MFCD30497694
  • Cas Number : 2059937-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321858
Availability details for
Quantity
  • Availability
  • Add to Cart