ENA638587234 Enamine

1-bromo-4-(trifluoromethoxy)-2-(trifluoromethyl)benzene

  • MDL number MFCD30497699
  • Cas Number : 2029355-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321969
Availability details for
Quantity
  • Availability
  • Add to Cart