ENA638587251 Enamine

3-methyl-5-[3-(trifluoromethyl)pyrrolidin-3-yl]-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD30497725
  • Cas Number : 2060039-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-323273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-323273
Availability details for
Quantity
  • Availability
  • Add to Cart