ENA638587270 Enamine

1-(4-methoxypyrrolidin-3-yl)-3-(propan-2-yl)urea hydrochloride

  • MDL number MFCD30497759
  • Cas Number : 2059975-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331434
Availability details for
Quantity
  • Availability
  • Add to Cart