ENA638587276 Enamine

1-[(benzyloxy)carbonyl]-3,3-dimethylazetidine-2-carboxylic acid

  • MDL number MFCD30497776
  • Cas Number : 2059937-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331506
Availability details for
Quantity
  • Availability
  • Add to Cart