ENA638587296 Enamine

4-(2,2-dimethylmorpholin-4-yl)benzoic acid hydrochloride

  • MDL number MFCD30497799
  • Cas Number : 2059975-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331657
Availability details for
Quantity
  • Availability
  • Add to Cart