ENA638587298 Enamine

3-(aminomethyl)-6-tert-butyl-1,2-dihydropyridin-2-one hydrochloride

  • MDL number MFCD30497801
  • Cas Number : 2059949-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331666
Availability details for
Quantity
  • Availability
  • Add to Cart