ENA638587315 Enamine

5-(chloromethyl)-5H,6H,7H-pyrrolo[1,2-b][1,2,4]triazole hydrochloride

  • MDL number MFCD30497825
  • Cas Number : 2059937-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331840
Availability details for
Quantity
  • Availability
  • Add to Cart