ENA638587319 Enamine

2-methanesulfonylcyclopropane-1,1-dicarboxylic acid

  • MDL number MFCD30497829
  • Cas Number : 2059948-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331854
Availability details for
Quantity
  • Availability
  • Add to Cart