ENA638587322 Enamine

N-[(3-amino-5-fluorophenyl)methyl]-2,2,2-trifluoroacetamide

  • MDL number MFCD30497837
  • Cas Number : 2059949-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331916
Availability details for
Quantity
  • Availability
  • Add to Cart