ENA638587330 Enamine

[5-(oxan-3-yl)-1,3,4-oxadiazol-2-yl]methanamine; oxalic acid

  • MDL number MFCD30497847
  • Cas Number : 2059949-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HOOCCOOH
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331982
Availability details for
Quantity
  • Availability
  • Add to Cart