ENA638587337 Enamine

2-(6-methoxy-7-methyl-1-benzofuran-3-yl)acetic acid

  • MDL number MFCD30497859
  • Cas Number : 2060029-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332039
Availability details for
Quantity
  • Availability
  • Add to Cart