ENA638587366 Enamine

3-{[(1r,4r)-4-aminocyclohexyl]amino}-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD30497903
  • Cas Number : 2059914-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332448
Availability details for
Quantity
  • Availability
  • Add to Cart