ENA638587369 Enamine

3-{5,7-dimethyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidin-6-yl}propanoic acid

  • MDL number MFCD30535667
  • Cas Number : 2059950-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332500
Availability details for
Quantity
  • Availability
  • Add to Cart