ENA638587383 Enamine

tert-butyl 5-(1-methyl-1H-pyrazol-4-yl)piperidine-3-carboxylate

  • MDL number MFCD30535669
  • Cas Number : 2059938-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332681
Availability details for
Quantity
  • Availability
  • Add to Cart