ENA638587389 Enamine

methyl 3-(2-bromophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoate

  • MDL number MFCD30497933
  • Cas Number : 1822499-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332729
Availability details for
Quantity
  • Availability
  • Add to Cart