ENA638587443 Enamine

7-bromo-3,3-dimethyl-2,3-dihydro-1H-indole

  • MDL number MFCD11847451
  • Cas Number : 1260675-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322344
Availability details for
Quantity
  • Availability
  • Add to Cart