ENA638587449 Enamine

5-fluoro-N-(2-methoxyethyl)pyridin-2-amine

  • MDL number MFCD14621287
  • Cas Number : 1248197-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322419
Availability details for
Quantity
  • Availability
  • Add to Cart