ENA638587482 Enamine

methyl 3-cyclobutyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD28097969
  • Cas Number : 2156772-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322766
Availability details for
Quantity
  • Availability
  • Add to Cart