ENA638587484 Enamine

3-(trifluoromethyl)-5H,6H,7H,8H-imidazo[1,5-a]pyridine-1-carboxylic acid

  • MDL number MFCD29079553
  • Cas Number : 1780407-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-318037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-318037
Availability details for
Quantity
  • Availability
  • Add to Cart