ENA638587588 Enamine

1-(3,5-dichlorophenoxy)-3-(methylamino)propan-2-ol

  • MDL number MFCD00826772
  • Cas Number : 350712-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331425
Availability details for
Quantity
  • Availability
  • Add to Cart