ENA638587592 Enamine

4-(4-fluorophenyl)-5-methyl-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD11220487
  • Cas Number : 1095231-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331430
Availability details for
Quantity
  • Availability
  • Add to Cart