ENA638587604 Enamine

7,7-dimethyl-1-phenyl-1H,4H,5H,6H,7H,8H-pyrazolo[4,3-c]azepine dihydrochloride

  • MDL number MFCD30497765
  • Cas Number : 2060036-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331459
Availability details for
Quantity
  • Availability
  • Add to Cart