ENA638587608 Enamine

2-chloro-1,3-benzothiazole-6-sulfonamide

  • MDL number MFCD28670610
  • Cas Number : 6608-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331468
Availability details for
Quantity
  • Availability
  • Add to Cart