ENA638587611 Enamine

2,3-dichloro-N-(cyclopropylmethyl)aniline hydrochloride

  • MDL number MFCD30497767
  • Cas Number : 2060021-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331472
Availability details for
Quantity
  • Availability
  • Add to Cart