ENA638587618 Enamine

5-(chloromethyl)-3,4-dihydro-1H-2-benzopyran

  • MDL number MFCD20637534
  • Cas Number : 1032590-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331482

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-331482
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart