ENA638587624 Enamine

1-(cyclobutylmethyl)-1H-pyrazole

  • MDL number MFCD20491913
  • Cas Number : 725746-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331496
Availability details for
Quantity
  • Availability
  • Add to Cart