ENA638587633 Enamine

N-ethyl-2-fluoro-3-methylaniline

  • MDL number MFCD29978784
  • Cas Number : 1696541-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331510
Availability details for
Quantity
  • Availability
  • Add to Cart