ENA638587642 Enamine

2-ethyl-5H,6H,7H,8H-pyrido[4,3-d]pyrimidine

  • MDL number MFCD22397549
  • Cas Number : 1598036-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331543
Availability details for
Quantity
  • Availability
  • Add to Cart