ENA638587670 Enamine

potassium 2-{2-[(2-cyclopropylethyl)amino]-1,3-thiazol-4-yl}acetate

  • MDL number MFCD30497789
  • Cas Number : 2059971-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331590
Availability details for
Quantity
  • Availability
  • Add to Cart