ENA638587676 Enamine

(1r,4r)-4-[(pyrazin-2-yl)amino]cyclohexan-1-ol

  • MDL number MFCD18837371
  • Cas Number : 1289385-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331597
Availability details for
Quantity
  • Availability
  • Add to Cart