ENA638587695 Enamine

2-[1-(3,5-dimethylphenyl)-2,5-dimethyl-1H-pyrrol-3-yl]acetic acid

  • MDL number MFCD01688619
  • Cas Number : 42866-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-318246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-318246
Availability details for
Quantity
  • Availability
  • Add to Cart