ENA638587728 Enamine

ethyl 6-bromopyrazolo[1,5-a]pyrimidine-2-carboxylate

  • MDL number MFCD11616111
  • Cas Number : 1005209-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331710
Availability details for
Quantity
  • Availability
  • Add to Cart