ENA638587730 Enamine

(1s,3s)-3-(phenylsulfanyl)cyclobutan-1-ol

  • MDL number MFCD30497814
  • Cas Number : 2059910-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331713
Availability details for
Quantity
  • Availability
  • Add to Cart