ENA638587743 Enamine

2-[2-(3,3,3-trifluoropropyl)-1,3-thiazol-4-yl]acetic acid hydrochloride

  • MDL number MFCD30497819
  • Cas Number : 2060060-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331758
Availability details for
Quantity
  • Availability
  • Add to Cart