ENA638587772 Enamine

3-(3-chlorophenyl)prop-2-ynenitrile

  • MDL number MFCD26636680
  • Cas Number : 935528-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331834
Availability details for
Quantity
  • Availability
  • Add to Cart