ENA638587785 Enamine

3-oxo-2,3-dihydroisoquinoline-8-carbonitrile

  • MDL number MFCD18803148
  • Cas Number : 1175272-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-318367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-318367
Availability details for
Quantity
  • Availability
  • Add to Cart