ENA638587788 Enamine

6-aminoquinolin-4-ol

  • MDL number MFCD09261102
  • Cas Number : 56717-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331853
Availability details for
Quantity
  • Availability
  • Add to Cart