ENA638587797 Enamine

2-(5-methylthiophen-2-yl)aniline hydrochloride

  • MDL number MFCD30497830
  • Cas Number : 2060047-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331884
Availability details for
Quantity
  • Availability
  • Add to Cart