ENA638587809 Enamine

[4-(2,2,2-trifluoroethyl)phenyl]methanol

  • MDL number MFCD28522957
  • Cas Number : 1108151-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331906
Availability details for
Quantity
  • Availability
  • Add to Cart