ENA638587811 Enamine

tert-butyl N-[3-(aminomethyl)-5-fluorophenyl]carbamate

  • MDL number MFCD30497836
  • Cas Number : 1893851-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331915
Availability details for
Quantity
  • Availability
  • Add to Cart