ENA638587825 Enamine

N-butyl-6-chloropyridin-3-amine

  • MDL number MFCD24255440
  • Cas Number : 120739-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-331965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-331965
Availability details for
Quantity
  • Availability
  • Add to Cart