ENA638587936 Enamine

ethyl 4-hydroxy-5-methyl-1,3-thiazole-2-carboxylate

  • MDL number MFCD11112110
  • Cas Number : 133833-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332239
Availability details for
Quantity
  • Availability
  • Add to Cart