ENA638587939 Enamine

1-cyclopentylpropan-2-amine

  • MDL number MFCD00190668
  • Cas Number : 105-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332243
Availability details for
Quantity
  • Availability
  • Add to Cart