ENA638587976 Enamine

1-(5-aminopyridin-2-yl)-3-(methoxymethyl)-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD16733221
  • Cas Number : 1249072-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332349
Availability details for
Quantity
  • Availability
  • Add to Cart