ENA638588010 Enamine

4-(2,6-dimethylmorpholin-4-yl)benzoic acid

  • MDL number MFCD11156519
  • Cas Number : 1021243-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332446
Availability details for
Quantity
  • Availability
  • Add to Cart