ENA638588022 Enamine

2-cyclobutanecarbonyloxolane

  • MDL number MFCD28646198
  • Cas Number : 1600972-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332475
Availability details for
Quantity
  • Availability
  • Add to Cart