ENA638588037 Enamine

5-nitro-2-(1H-1,2,3,4-tetrazol-1-yl)benzoic acid

  • MDL number MFCD09047082
  • Cas Number : 926199-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332512

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332512
Availability details for
Quantity
  • Availability
  • Add to Cart