ENA638588044 Enamine

3-cyclobutanecarbonyloxolane

  • MDL number MFCD21117519
  • Cas Number : 1495485-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332528
Availability details for
Quantity
  • Availability
  • Add to Cart