ENA638588081 Enamine

methyl 2-(3-oxothiomorpholin-2-yl)acetate

  • MDL number MFCD02669633
  • Cas Number : 73477-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332632
Availability details for
Quantity
  • Availability
  • Add to Cart